MilliporeSigma
Age-Related Autonomous Aldosteronism.
Circulation (2017-06-02)
Kazutaka Nanba, Anand Vaidya, Gordon H Williams, Isabel Zheng, Tobias Else, William E Rainey
ABSTRACT
Both aging and inappropriate secretion of aldosterone increase the risk for developing cardiovascular disease; however, the influence of aging on aldosterone secretion and physiology is not well understood. The relationship between age and adrenal aldosterone synthase (CYP11B2) expression was evaluated in 127 normal adrenal glands from deceased kidney donors (age, 9 months to 68 years). Following immunohistochemistry, CYP11B2-expressing area and areas of abnormal foci of CYP11B2-expressing cells, called aldosterone-producing cell clusters, were analyzed. In a separate ancillary clinical study of 677 participants without primary aldosteronism, who were studied on both high and restricted sodium diets (age, 18-71 years), we used multivariable linear regression to assess the independent associations between age and renin-angiotensin-aldosterone system physiology. In adrenal tissue, the total CYP11B2-expressing area was negatively correlated with age (r=-0.431, P<0.0001), whereas the total aldosterone-producing cell cluster area was positively correlated with age (r=0.390, P<0.0001). The integrated ratio of aldosterone-producing cell cluster to CYP11B2-expressing area was most strongly and positively correlated with age (r=0.587, P<0.0001). When participants in the clinical study were maintained on a high sodium balance, renin activity progressively declined with older age, whereas serum and urinary aldosterone did not significantly decline. Correspondingly, the aldosterone-to-renin ratio was positively and independently associated with older age (adjusted β=+5.54 ng/dL per ng/mL per hour per 10 years, P<0.001). In contrast, when participants were assessed under sodium-restricted conditions, physiological stimulation of aldosterone was blunted with older age (β=-4.6 ng/dL per 10 years, P<0.0001). Aging is associated with a pattern of decreased normal zona glomerulosa CYP11B2 expression and increased aldosterone-producing cell cluster expression. This histopathologic finding parallels an age-related autonomous aldosteronism and abnormal aldosterone physiology that provides 1 potential explanation for age-related cardiovascular risk.
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
Anti-CYP11B2 Antibody, clone 41-17B, clone 41-17B, from mouse