MilliporeSigma
  • SIX2 haploinsufficiency causes conductive hearing loss with ptosis in humans.
SIX2 haploinsufficiency causes conductive hearing loss with ptosis in humans.
Journal of human genetics (2016-07-08)
Jing Guan, Dayong Wang, Wenjian Cao, Yali Zhao, Renqian Du, Hu Yuan, Qiong Liu, Lan Lan, Liang Zong, Ju Yang, Zifang Yin, Bing Han, Feng Zhang, Qiuju Wang
ABSTRACT
The ossicles represent one of the most fundamental morphological features in evolutionary biology of the mammalians. The mobile ossicular morphology abnormalities result in the severe conductive hearing loss. Development and patterning of the middle ear malformation depend on genetic and environmental causes. However, the genetic basis for the risk of congenital ossicle malformation is poorly understood. We show here nine affected individuals in a Chinese pedigree who had bilateral conductive hearing loss with ptosis. We performed whole-genome sequencing and array comparative genomic hybridization (CGH) analysis on DNA samples from the Chinese pedigree. We confirmed the presence of a novel 60 kb heterozygous deletion in size, encompassing SIX2 in our family. Mutation screening in 169 sporadic cases with external ear and middle ear malformations identified no pathogenic variant or polymorphism. We suggest SIX2 haploinsufficiency as a potential congenital factor could be attributed to developmental malformation of the middle ear ossicles and upper eyelid. To the best of our knowledge, this is the first report to provide a description of copy number variation in the SIX2 gene resulting in syndromic conductive hearing loss.