MilliporeSigma
  • Polychlorinated biphenyls disrupt hepatic epidermal growth factor receptor signaling.
Polychlorinated biphenyls disrupt hepatic epidermal growth factor receptor signaling.
Xenobiotica; the fate of foreign compounds in biological systems (2016-07-28)
Josiah E Hardesty, Banrida Wahlang, K Cameron Falkner, Heather B Clair, Barbara J Clark, Brian P Ceresa, Russell A Prough, Matthew C Cave
ABSTRACT
1. Polychlorinated biphenyls (PCBs) are persistent environmental pollutants that disrupt hepatic xenobiotic and intermediary metabolism, leading to metabolic syndrome and nonalcoholic steatohepatitis (NASH). 2. Since phenobarbital indirectly activates Constitutive Androstane Receptor (CAR) by antagonizing growth factor binding to the epidermal growth factor receptor (EGFR), we hypothesized that PCBs may also diminish EGFR signaling. 3. The effects of the PCB mixture Aroclor 1260 on the protein phosphorylation cascade triggered by EGFR activation were determined in murine (in vitro and in vivo) and human models (in vitro). EGFR tyrosine residue phosphorylation was decreased by PCBs in all models tested. 4. The IC
Sigma-Aldrich
TCPOBOP, ≥98% (HPLC), solid
Sigma-Aldrich
EGFR Inhibitor, The EGFR Inhibitor, also referenced under CAS 879127-07-8, controls the biological activity of EGFR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.