MilliporeSigma
  • Neuroimmune-Glia Interactions in the Sensory Ganglia Account for the Development of Acute Herpetic Neuralgia.
Neuroimmune-Glia Interactions in the Sensory Ganglia Account for the Development of Acute Herpetic Neuralgia.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-06-04)
Jaqueline R Silva, Alexandre H Lopes, Jhimmy Talbot, Nerry T Cecilio, Mateus F Rossato, Rangel L Silva, Guilherme R Souza, Cassia R Silva, Guilherme Lucas, Benedito A Fonseca, Eurico Arruda, Jose C Alves-Filho, Fernando Q Cunha, Thiago M Cunha
ABSTRACT
Herpetic neuralgia is the most important symptom of herpes zoster disease, which is caused by