Merck
  • Osthole improves function of periodontitis periodontal ligament stem cells via epigenetic modification in cell sheets engineering.
Osthole improves function of periodontitis periodontal ligament stem cells via epigenetic modification in cell sheets engineering.
Scientific reports (2017-07-14)
Jin Sun, Zhiwei Dong, Yang Zhang, Xiaoning He, Dongdong Fei, Fang Jin, Lin Yuan, Bei Li, Yan Jin
ABSTRACT
Inflammatory microenvironment causes the change of epigenetic modification in periodontal ligament stem cells derived from periodontitis tissues (P-PDLSCs), which results in defective osteogenic differentiation compared to cells from healthy tissues. It's urgent to explore therapeutic strategies aimed at epigenetic targets associated with the regenerative ability of PDLSCs. Osthole, a small-molecule compound extracted from Chinese herbs, has been documented to promote osteogenesis and cell sheets formation of healthy PDLSCs. However, whether osthole shows same effect on P-PDLSCs and the mechanism of promotive effect is still unknown. The purpose of this study was to determine whether Osthole could restore defective osteogenic differentiation of P-PDLSCs via epigenetic modification. We demonstrated that 10