MilliporeSigma
Quantitative analysis of protein interaction network dynamics in yeast.
Molecular systems biology (2017-07-15)
Albi Celaj, Ulrich Schlecht, Justin D Smith, Weihong Xu, Sundari Suresh, Molly Miranda, Ana Maria Aparicio, Michael Proctor, Ronald W Davis, Frederick P Roth, Robert P St Onge
ABSTRACT
Many cellular functions are mediated by protein-protein interaction networks, which are environment dependent. However, systematic measurement of interactions in diverse environments is required to better understand the relative importance of different mechanisms underlying network dynamics. To investigate environment-dependent protein complex dynamics, we used a DNA-barcode-based multiplexed protein interaction assay in
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis