MilliporeSigma
  • Folate Metabolism Regulates Oligodendrocyte Survival and Differentiation by Modulating AMPKα Activity.
Folate Metabolism Regulates Oligodendrocyte Survival and Differentiation by Modulating AMPKα Activity.
Scientific reports (2017-05-13)
Qinjie Weng, Jiajia Wang, Jiaying Wang, Biqin Tan, Jing Wang, Haibo Wang, Tao Zheng, Q Richard Lu, Bo Yang, Qiaojun He
ABSTRACT
Folate, an essential micronutrient, is a critical cofactor in one-carbon metabolism for many cellular pathways including DNA synthesis, metabolism and maintenance. Folate deficiency has been associated with an increased risk of neurological disease, cancer and cognitive dysfunction. Dihydrofolate reductase (DHFR) is a key enzyme to regulate folate metabolism, however folate/DHFR activity in oligodendrocyte development has not been fully understood. Here we show that folate enhances oligodendrocyte maturation both in vitro and in vivo, which is accompanied with upregulation of oligodendrocyte-specific DHFR expression. On the other hand, pharmacological inhibition of DHFR by methotrexate (MTX) causes severe defects in oligodendrocyte survival and differentiation, which could be reversed by folate intake. We further demonstrate that folate activates a metabolic regulator AMPKα to promote oligodendrocyte survival and differentiation. Moreover, activation of AMPKα partially rescues oligodendrocyte defects caused by DHFR-inhibition both in vitro and in vivo. Taken together, these findings identify a previously uncharacterized role of folate/DHFR/AMPKα axis in regulating oligodendrocyte survival and myelination during CNS development.
Sigma-Aldrich
Anti-Myelin Proteolipid Protein Antibody, CT, clone PLPC1, clone PLPC1, Chemicon®, from mouse
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, 10 ×, With 1000 mg/L glucose (1x), without L-glutamine, sodium bicarbonate, and folic acid, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse