MilliporeSigma
  • The E3 Ligase CHIP Mediates p21 Degradation to Maintain Radioresistance.
The E3 Ligase CHIP Mediates p21 Degradation to Maintain Radioresistance.
Molecular cancer research : MCR (2017-02-25)
Kuntal Biswas, Sukumar Sarkar, Kangping Du, David L Brautigan, Tarek Abbas, James M Larner
ABSTRACT
Lung cancer resists radiotherapy, making it one of the deadliest forms of cancer. Here, we show that human lung cancer cell lines can be rendered sensitive to ionizing radiation (IR) by RNAi knockdown of C-terminus of Hsc70-interacting protein (CHIP/STUB1), a U-box-type E3 ubiquitin ligase that targets a number of stress-induced proteins. Mechanistically, ubiquitin-dependent degradation of the cyclin-dependent kinase (CDK) inhibitor, p21 protein, is reduced by CHIP knockdown, leading to enhanced senescence of cells in response to exposure to IR. Cellular senescence and sensitivity to IR is prevented by CRISPR/Cas9-mediated deletion of the p21 gene (