MilliporeSigma
Ubiquitination and dynactin regulate TMEPAI lysosomal trafficking.
Scientific reports (2017-02-22)
Shenheng Luo, Lei Jing, Tian Zhao, Yuyin Li, Zhenxing Liu, Aipo Diao
ABSTRACT
The transmembrane prostate androgen-induced protein (TMEPAI) has been reported to be elevated in various tumor cells, is localized to the lysosome and promotes lysosome stability. The molecular mechanism of TMEPAI trafficking however to the lysosome is unknown. Here we report that clathrin and CI-M6PR mediate TMEPAI transport from the Golgi directly into the endo-lysosomal pathway. TMEPAI is ubiquitinated at its C-terminal region and ubiquitination modification of TMEPAI is a signal for its lysosomal trafficking. Moreover, TMEPAI binds the ubiquitin binding proteins Hrs and STAM which is required for its lysosomal transport. In addition, TMEPAI interacts with the dynactin pointed-end complex subunits dynactin 5 and dynactin 6. The aa 132-155 domain is essential for specific TMEPAI binding and deletion of this binding site leads to mis-trafficking of TMEPAI to the plasma membrane. These results reveal the pathway and mechanism regulating transport of TMEPAI to the lysosome, which helps to further understand the role of TMEPAI in tumorigenesis.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes