MilliporeSigma
  • Long non-coding RNA H19 enhances cell proliferation and anchorage-independent growth of cervical cancer cell lines.
Long non-coding RNA H19 enhances cell proliferation and anchorage-independent growth of cervical cancer cell lines.
Experimental biology and medicine (Maywood, N.J.) (2016-09-17)
Tawin Iempridee
ABSTRACT
Long non-coding RNA H19 is aberrantly expressed in multiple malignancies and its expression levels correlate with recurrence, metastasis, and patient survival. Despite numerous reports documenting the role of H19 in carcinogenesis, its contribution to cervical cancer development is still largely unknown. In this study, I observed that H19 expression was elevated in cervical cancer cell lines and could be detected in extracellular vesicles in the culture medium. In addition, I demonstrated, by overexpression and knockdown experiments, that H19 promoted cell proliferation and multicellular tumor spheroid formation without significantly affecting apoptosis and cell migration. Finally, treatment with transforming growth factor beta and hypoxia-mimetic CoCl
Sigma-Aldrich
Doxycycline hydrochloride
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture