MilliporeSigma
  • A simple strategy for extracellular production of CotA laccase in Escherichia coli and decolorization of simulated textile effluent by recombinant laccase.
A simple strategy for extracellular production of CotA laccase in Escherichia coli and decolorization of simulated textile effluent by recombinant laccase.
Applied microbiology and biotechnology (2016-10-16)
Tian-Nyu Wang, Min Zhao
ABSTRACT
Laccases are green oxidases with a number of potential industrial applications. In this study, recombinant Bacillus subtilis CotA laccase was secreted by Escherichia coli via both the α-hemolysin secretion system and the YebF secretion system after microaerobic induction. Meanwhile, we discovered a much simpler approach for extracellular production of recombinant CotA laccase from E. coli, involving alternation of induction conditions to release recombinant CotA following intracellular expression. By optimizing the induction parameters, the extracellular yield of recombinant CotA laccase was improved from 157.4 to 2401.3 U/L after 24 h of induction. This strategy could be suitable for large-scale production of CotA laccase for industrial use. Recombinant CotA laccase was purified by Ni
Sigma-Aldrich
2-Hydroxyacetophenone, 98%