MilliporeSigma
  • Oxidative Stress and Nuclear Factor κB (NF-κB) Increase Peritoneal Filtration and Contribute to Ascites Formation in Nephrotic Syndrome.
Oxidative Stress and Nuclear Factor κB (NF-κB) Increase Peritoneal Filtration and Contribute to Ascites Formation in Nephrotic Syndrome.
The Journal of biological chemistry (2016-04-02)
Khalil Udwan, Gaëlle Brideau, Marc Fila, Aurélie Edwards, Bruno Vogt, Alain Doucet
ABSTRACT
Water accumulation in the interstitium (edema) and the peritoneum (ascites) of nephrotic patients is classically thought to stem from the prevailing low plasma albumin concentration and the decreased transcapillary oncotic pressure gradient. However, several clinical and experimental observations suggest that it might also stem from changes in capillary permeability. We addressed this hypothesis by studying the peritoneum permeability of rats with puromycin aminonucleoside-induced nephrotic syndrome. The peritoneum of puromycin aminonucleoside rats displayed an increase in the water filtration coefficient of paracellular and transcellular pathways, and a decrease in the reflection coefficient to proteins. It also displayed oxidative stress and subsequent activation of NF-κB. Scavenging of reactive oxygen species and inhibition of NF-κB prevented the changes in the water permeability and reflection coefficient to proteins and reduced the volume of ascites by over 50%. Changes in water permeability were associated with the overexpression of the water channel aquaporin 1, which was prevented by reactive oxygen species scavenging and inhibition of NF-κB. In conclusion, nephrotic syndrome is associated with an increased filtration coefficient of the peritoneum and a decreased reflection coefficient to proteins. These changes, which account for over half of ascite volume, are triggered by oxidative stress and subsequent activation of NF-κB.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Nitrotyrosine Antibody, Upstate®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Puromycin aminonucleoside Ready Made Solution, 10 mg/mL in water