MilliporeSigma
  • High content image analysis identifies novel regulators of synaptogenesis in a high-throughput RNAi screen of primary neurons.
High content image analysis identifies novel regulators of synaptogenesis in a high-throughput RNAi screen of primary neurons.
PloS one (2014-03-19)
Thomas J F Nieland, David J Logan, Jessica Saulnier, Daniel Lam, Caroline Johnson, David E Root, Anne E Carpenter, Bernardo L Sabatini
ABSTRACT
The formation of synapses, the specialized points of chemical communication between neurons, is a highly regulated developmental process fundamental to establishing normal brain circuitry. Perturbations of synapse formation and function causally contribute to human developmental and degenerative neuropsychiatric disorders, such as Alzheimer's disease, intellectual disability, and autism spectrum disorders. Many genes controlling synaptogenesis have been identified, but lack of facile experimental systems has made systematic discovery of regulators of synaptogenesis challenging. Thus, we created a high-throughput platform to study excitatory and inhibitory synapse development in primary neuronal cultures and used a lentiviral RNA interference library to identify novel regulators of synapse formation. This methodology is broadly applicable for high-throughput screening of genes and drugs that may rescue or improve synaptic dysfunction associated with cognitive function and neurological disorders.
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Synapsin I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse