MilliporeSigma
  • 5-Fluorouracil affects assembly of stress granules based on RNA incorporation.
5-Fluorouracil affects assembly of stress granules based on RNA incorporation.
Nucleic acids research (2014-04-15)
Christian Kaehler, Jörg Isensee, Tim Hucho, Hans Lehrach, Sylvia Krobitsch
ABSTRACT
The antimetabolite 5-fluorouracil is a widely used chemotherapeutic for the treatment of several solid cancers. However, resistance to 5-fluorouracil remains a major drawback in its clinical use. In this study we report that treatment of HeLa cells with 5-fluorouracil resulted in de novo assembly of stress granules. Moreover, we revealed that stress granule assembly under stress conditions as well as disassembly is altered in cells treated with 5-fluorouracil. Notably, we discovered that RACK1, a protein mediating cell survival and apoptosis, is a component of 5-fluorouracil-induced stress granules. To explore the mode of action of 5-fluorouracil accountable for de novo stress granule assembly, we analyzed 5-fluorouracil metabolites and noticed that stress granule assembly is caused by RNA, not DNA incorporating 5-fluorouracil metabolites. Interestingly, we observed that other RNA incorporating drugs also cause assembly of stress granules. Thus, our results suggest that incorporation of chemotherapeutics into RNA may result in stress granule assembly with potential significance in chemoresistance.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody