MilliporeSigma
High-grade myofibroblastic sarcoma of inguinal region in a dog.
The Journal of veterinary medical science (2011-12-14)
Yuri Hojo, Takuma Tsuchiya, Ayako Shiraki, Kazuhiko Suzuki, Makoto Shibutani, Kunitoshi Mitsumori
ABSTRACT
A subcutaneous tumor in the left inguinal region was present in an 11-year-old female bloodhound. Histopathologically, the tumor showed invasive growth and extensive necroses, and it was composed of spindle-shaped, elongated, and stellate neoplastic cells accompanied by occasional giant cells arranged in fascicular, herringbone, or irregular storiform patterns with abundant production of collagen fibers. The cytoplasm of most tumor cells was positive for vimentin, alpha-smooth muscle actin, and calponin, but was negative for desmin, smoothelin, and S-100. Furthermore, most of the tumor cells were negative for Iba1 while some tumor cells were weakly positive. Thus, this tumor was diagnosed as a high-grade myofibroblastic sarcoma according to the diagnostic criteria for human myofibroblastic sarcomas.
Sigma-Aldrich
Monoclonal Anti-Calponin antibody produced in mouse, clone hCP, ascites fluid
Sigma-Aldrich
Anti-Smoothelin Antibody, clone R4A, clone R4A, Chemicon®, from mouse