MilliporeSigma
  • Human Immunodeficiency Virus Type 1 Nef Inhibits Autophagy through Transcription Factor EB Sequestration.
Human Immunodeficiency Virus Type 1 Nef Inhibits Autophagy through Transcription Factor EB Sequestration.
PLoS pathogens (2015-06-27)
Grant R Campbell, Pratima Rawat, Rachel S Bruckman, Stephen A Spector
ABSTRACT
HIV Nef acts as an anti-autophagic maturation factor through interaction with beclin-1 (BECN1). We report that exposure of macrophages to infectious or non-infectious purified HIV induces toll-like receptor 8 (TLR8) and BECN1 dependent dephosphorylation and nuclear translocation of TFEB and that this correlates with an increase in autophagy markers. RNA interference for ATG13, TFEB, TLR8, or BECN1 inhibits this HIV-induced autophagy. However, once HIV establishes a productive infection, TFEB phosphorylation and cytoplasmic sequestration are increased resulting in decreased autophagy markers. Moreover, by 7 d post-infection, autophagy levels are similar to mock infected controls. Conversely, although Nef deleted HIV similarly induces TFEB dephosphorylation and nuclear localization, and increases autophagy, these levels remain elevated during continued productive infection. Thus, the interaction between HIV and TLR8 serves as a signal for autophagy induction that is dependent upon the dephosphorylation and nuclear translocation of TFEB. During permissive infection, Nef binds BECN1 resulting in mammalian target of rapamycin (MTOR) activation, TFEB phosphorylation and cytosolic sequestration, and the inhibition of autophagy. To our knowledge, this is the first report of a virus modulating TFEB localization and helps to explain how HIV modulates autophagy to promote its own replication and cell survival.
Sigma-Aldrich
Monoclonal Anti-TLR8 antibody produced in mouse, clone 4C6, purified immunoglobulin, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid