MilliporeSigma
  • Sperm abnormalities induced by pre-pubertal exposure to cyclophosphamide are effectively mitigated by Moringa oleifera leaf extract.
Sperm abnormalities induced by pre-pubertal exposure to cyclophosphamide are effectively mitigated by Moringa oleifera leaf extract.
Andrologia (2015-04-24)
G Nayak, A Vadinkar, S Nair, S G Kalthur, A S D'Souza, P K Shetty, S Mutalik, M M Shetty, G Kalthur, S K Adiga
ABSTRACT
Moringa oleifera L. is a medicinal plant with potential antioxidant property. This study was aimed at investigating the chemoprotective effect of Moringa oleifera leaf extract (MOE) on cyclophosphamide (CP)-induced testicular toxicity. Two-week-old male Swiss albino mice were intraperitoneally injected with phosphate-buffered saline, 50 mg kg(-1) of CP and 25 mg kg(-1) of MOE. In combination treatment, mice were injected with 25 mg kg(-1) of MOE 24 h prior to CP injection, 24 h prior and post-CP injection and 24 h post-CP injection for 5 consecutive days (10 mg kg(-1) ). Six weeks later, mice were sacrificed to assess epididymal sperm parameters. MOE alone did not have any significant effect on sperm parameters. However, acute injection of CP resulted in significant decline in motility (P < 0.001), increase in head abnormality (P < 0.01) and DNA damage (P < 0.05). Combining MOE with CP increased the sperm density, motility and reduced head defect and DNA damage, irrespective of the schedule and dosage of MOE. Administration of MOE prior to CP significantly elevated the level of superoxide dismutase and catalase with concomitant decrease in lipid peroxidation in the testicular tissue. In conclusion, MOE may have potential benefit in reducing the loss of male gonadal function following chemotherapy.
Sigma-Aldrich
Medium 199, With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra