MilliporeSigma
  • Epstein-Barr virus transcription factor Zta acts through distal regulatory elements to directly control cellular gene expression.
Epstein-Barr virus transcription factor Zta acts through distal regulatory elements to directly control cellular gene expression.
Nucleic acids research (2015-03-18)
Sharada Ramasubramanyan, Kay Osborn, Rajaei Al-Mohammad, Ijiel B Naranjo Perez-Fernandez, Jianmin Zuo, Nicolae Balan, Anja Godfrey, Harshil Patel, Gordon Peters, Martin Rowe, Richard G Jenner, Alison J Sinclair
ABSTRACT
Lytic replication of the human gamma herpes virus Epstein-Barr virus (EBV) is an essential prerequisite for the spread of the virus. Differential regulation of a limited number of cellular genes has been reported in B-cells during the viral lytic replication cycle. We asked whether a viral bZIP transcription factor, Zta (BZLF1, ZEBRA, EB1), drives some of these changes. Using genome-wide chromatin immunoprecipitation coupled to next-generation DNA sequencing (ChIP-seq) we established a map of Zta interactions across the human genome. Using sensitive transcriptome analyses we identified 2263 cellular genes whose expression is significantly changed during the EBV lytic replication cycle. Zta binds 278 of the regulated genes and the distribution of binding sites shows that Zta binds mostly to sites that are distal to transcription start sites. This differs from the prevailing view that Zta activates viral genes by binding exclusively at promoter elements. We show that a synthetic Zta binding element confers Zta regulation at a distance and that distal Zta binding sites from cellular genes can confer Zta-mediated regulation on a heterologous promoter. This leads us to propose that Zta directly reprograms the expression of cellular genes through distal elements.
Sigma-Aldrich
Anti-EBV VCA gp125 Antibody, clone L2, clone L2, Chemicon®, from mouse
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture