MilliporeSigma
  • Expression in brain of a messenger RNA encoding a novel neuropeptide homologous to calcitonin gene-related peptide.
Expression in brain of a messenger RNA encoding a novel neuropeptide homologous to calcitonin gene-related peptide.
Science (New York, N.Y.) (1985-09-13)
S G Amara, J L Arriza, S E Leff, L W Swanson, R M Evans, M G Rosenfeld
ABSTRACT
As a consequence of alternative RNA processing events, a single rat gene can generate messenger RNA's (mRNA's) encoding either calcitonin or a neuropeptide referred to as alpha-type calcitonin gene-related peptide (alpha-CGRP). An mRNA product of a related gene has been identified in rat brain and thyroid encoding the protein precursor of a peptide differing from alpha-CGRP by only a single amino acid. The RNA encoding this peptide, which is referred to as beta-CGRP, appears to be the only mature transcript of the beta-CGRP gene. Hybridization histochemistry reveals a similar distribution of alpha- and beta-CGRP mRNA's, but their relative levels of expression vary in different cranial nerve nuclei. Thus beta-CGRP is a new member of a family of related genes with potential functions in regulating the transduction of sensory and motor information.