MilliporeSigma
  • KPT-330 inhibitor of XPO1-mediated nuclear export has anti-proliferative activity in hepatocellular carcinoma.
KPT-330 inhibitor of XPO1-mediated nuclear export has anti-proliferative activity in hepatocellular carcinoma.
Cancer chemotherapy and pharmacology (2014-07-18)
Yun Zheng, Sigal Gery, Haibo Sun, Sharon Shacham, Michael Kauffman, H Phillip Koeffler
ABSTRACT
Exportin-1 (XPO1, CRM1) mediates the nuclear export of several key growth regulatory and tumor suppressor proteins. Cancer cells often overexpress XPO1 resulting in cytoplasmic mislocalization and aberrant activity of its target proteins. Orally bioavailable selective inhibitors of nuclear export (SINE) that irreversibly bind to and inhibit the function of XPO1 have been recently developed. The aim of this study was to investigate the efficacy of the clinical staged, orally available, SINE compound, KPT-330 in hepatocellular carcinoma (HCC). In silico, meta-analysis showed that XPO1 is overexpressed in HCC. Six HCC cell lines were treated with KPT-330, and cell proliferation and expression of cell growth regulators were examined by cell proliferation assays and Western blot analysis, respectively. The in vivo anti-cancer activity of KPT-330 was examined in a HCC xenograft murine model. KPT-330 reduced the viability of HCC cell lines in vitro and this anti-proliferative effect was associated with cell cycle arrest and induction of apoptosis. The expression of the pro-apoptotic protein PUMA was markedly up-regulated by KPT-330. In addition, SINE treatment increased the expression of the tumor suppressor proteins p53 and p27, while it reduced the expression of HCC promoting proteins, c-Myc and c-Met. XPO1 levels itself were also down-regulated following KPT-330 treatment. Finally, a HCC xenograft murine model showed that treatment of mice with oral KPT-330 significantly inhibited tumor growth with little evidence of toxicity. Our results suggest that SINE compounds, such as KPT-330, are promising novel drugs for the targeted therapy of HCC.
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma