MilliporeSigma
  • Temsirolimus induces surfactant lipid accumulation and lung inflammation in mice.
Temsirolimus induces surfactant lipid accumulation and lung inflammation in mice.
American journal of physiology. Lung cellular and molecular physiology (2014-05-06)
Satoshi Washino, Hitoshi Ando, Kentarou Ushijima, Keiko Hosohata, Masafumi Kumazaki, Naoko Mato, Yukihiko Sugiyama, Yutaka Kobayashi, Akio Fujimura, Tatsuo Morita
ABSTRACT
Interstitial lung disease (ILD) is a well-known adverse effect of mammalian target of rapamycin (mTOR) inhibitors. However, it remains unknown how lung toxicities are induced by mTOR inhibitors. Here, we constructed a mouse model of mTOR inhibitor-induced ILD using temsirolimus and examined the pathogenesis of the disease. Male ICR mice were treated with an intraperitoneal injection of different doses of temsirolimus (3 or 30 mg·kg(-1)·wk(-1)) or vehicle. Temsirolimus treatment increased capillary-alveolar permeability and induced neutrophil infiltration and fibrinous exudate into the alveolar space, indicating alveolar epithelial and/or endothelial injury. It also induced macrophage depletion and the accumulation of excessive surfactant phospholipids and cholesterols. Alveolar macrophage depletion is thought to cause surfactant lipid accumulation. To further examine whether temsirolimus has cytotoxic and/or cytostatic effects on alveolar macrophages and alveolar epithelial cells, we performed in vitro experiments. Temsirolimus inhibited cell proliferation and viability in both alveolar macrophage and alveolar epithelial cells. Temsirolimus treatment caused some signs of pulmonary inflammation, including upregulated expression of several proinflammatory cytokines in both bronchoalveolar lavage cells and lung homogenates, and an increase in lymphocytes in the bronchoalveolar lavage fluid. These findings indicate that temsirolimus has the potential to induce alveolar epithelial injury and to deplete alveolar macrophages followed by surfactant lipid accumulation, resulting in pulmonary inflammation. This is the first study to focus on the pathogenesis of mTOR inhibitor-induced ILD using an animal model.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Temsirolimus, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)