MilliporeSigma
  • Erythropoietin exerts cell protective effect by activating PI3K/Akt and MAPK pathways in C6 Cells.
Erythropoietin exerts cell protective effect by activating PI3K/Akt and MAPK pathways in C6 Cells.
Neurological research (2014-02-12)
Min-Soo Kwon, Mi-Hee Kim, Seon-Hee Kim, Ki-Dae Park, Si-Hyung Yoo, Il-Ung Oh, Suenie Pak, Young-Jun Seo
ABSTRACT
Even though erythropoietin (EPO) is a neurotropic cytokine that is recognized widely for its role in the development, maintenance, protection, and repair of the nervous system, there are few reports concerning EPO-mediated influences on the glial cells in the central nervous system. In this study, we investigated anti-inflammatory and anti-apoptotic effects of EPO on C6 glioma cells (C6 cells). Erythropoietin did not attenuate inflammatory response, such as nitrite production, iNOS gene expression, and pro-inflammatory cytokines when LPS/TNF-alpha mixture was treated. However, EPO increased C6 cell viability by exerting cell protective effect against staurosporine stimulation. Erythropoietin increased the transient Akt expression at 30 minutes and induced the gradual elevation of ERK1/2 and p38 expression as time progressed. The cell protective effect of EPO was also significantly attenuated with pretreatment of specific PI3K, pERK1/2, or pP38 inhibitor. In summary, these results suggest that EPO may exert its cell protective functions via the direct cell protective activity rather than via its anti-inflammatory effect. Moreover, the PI3K/Akt and mitogen activated protein kinase (MAPK) pathways may be responsible for cell survival against cytotoxicity.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Supelco
Staurosporine, VETRANAL®, analytical standard