MilliporeSigma
  • High frequencies of polyfunctional CD8+ NK cells in chronic HIV-1 infection are associated with slower disease progression.
High frequencies of polyfunctional CD8+ NK cells in chronic HIV-1 infection are associated with slower disease progression.
Journal of virology (2014-08-15)
Fareed Ahmad, Henoch S Hong, Marc Jäckel, Alexandra Jablonka, I-Na Lu, Nupur Bhatnagar, Johanna M Eberhard, Benjamin A Bollmann, Matthias Ballmaier, Margot Zielinska-Skowronek, Reinhold E Schmidt, Dirk Meyer-Olson
ABSTRACT
Natural killer (NK) cells are effector and regulatory innate immune cells and play a critical role in the first line of defense against various viral infections. Although previous reports have indicated the vital contributions of NK cells to HIV-1 immune control, nongenetic NK cell parameters directly associated with slower disease progression have not been defined yet. In a longitudinal, retrospective study of 117 untreated HIV-infected subjects, we show that higher frequencies as well as the absolute numbers of CD8(+) CD3(-) lymphocytes are linked to delayed HIV-1 disease progression. We show that the majority of these cells are well-described blood NK cells. In a subsequent cross-sectional study, we demonstrate a significant loss of CD8(+) NK cells in untreated HIV-infected individuals, which correlated with HIV loads and inversely correlated with CD4(+) T cell counts. CD8(+) NK cells had modestly higher frequencies of CD57-expressing cells than CD8(-) cells, but CD8(+) and CD8(-) NK cells showed no differences in the expression of a number of activating and inhibiting NK cell receptors. However, CD8(+) NK cells exhibited a more functional profile, as detected by cytokine production and degranulation. We demonstrate that the frequency of highly functional CD8(+) NK cells is inversely associated with HIV-related disease markers and linked with delayed disease progression. These results thus indicate that CD8(+) NK cells represent a novel NK cell-derived, innate immune correlate with an improved clinical outcome in HIV infection.
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications