MilliporeSigma
  • Rearrangements of MYC gene facilitate risk stratification in diffuse large B-cell lymphoma patients treated with rituximab-CHOP.
Rearrangements of MYC gene facilitate risk stratification in diffuse large B-cell lymphoma patients treated with rituximab-CHOP.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2013-12-18)
Alexandar Tzankov, Zijun Y Xu-Monette, Marc Gerhard, Carlo Visco, Stephan Dirnhofer, Nora Gisin, Karen Dybkaer, Attilio Orazi, Govind Bhagat, Kristy L Richards, Eric D Hsi, William W L Choi, J Han van Krieken, Maurilio Ponzoni, Andrés J M Ferreri, Qing Ye, Jane N Winter, John P Farnen, Miguel A Piris, Michael B Møller, M James You, Timothy McDonnell, L Jeffrey Medeiros, Ken H Young
ABSTRACT
In order to address the debatable prognostic role of MYC rearrangements in diffuse large B-cell lymphoma patients treated with rituximab, cyclophosphamide, hydroxydaunorubicin, vincristine, and prednisone, we evaluated MYC rearrangements by fluorescence in situ hybridization in 563 cases using break-apart probes and IGH/MYC dual-fusion probes. Concurrent BCL2 and BCL6 aberrations were also assessed. Data were correlated with clinicopathological variables and prognostic parameters. MYC rearrangements were observed in 39/432 evaluable cases (9%), including 4 rearrangements detectable only with the dual-fusion probes, 15 detectable only with the break-apart probes and 20 detectable with both dual-fusion probes and break-apart probes. MYC rearrangements correlated with germinal center B-cell origin (P=0.02), MYC protein expression (P=0.032), and larger tumor mass size (P=0.0003). Patients with MYC rearrangements were more likely to be treatment resistant (P<0.0001). All types of MYC rearrangements were associated with poorer disease-specific survival, that is, 20/39 dead, median disease-specific survival 42 months, compared with 98/393 dead among the non-rearranged cases, median disease-specific survival not reached (P=0.0002). Cases with MYC rearrangements that overexpressed MYC protein were at risk with respect to disease-specific survival independent of the International Prognostic Index (P=0.046 and P<0.001, respectively). Presence of concurrent BCL2 aberrations but not of BCL6 aberrations was prognostically additive. Radiotherapy seemed to diminish the prognostic effects of MYC rearrangements in diffuse large B-cell lymphoma patients since only 2/10 irradiated patients with MYC rearrangements died of/with disease, compared with 16/28 non-irradiated patients with MYC rearrangements. We conclude that MYC rearrangements add prognostic information for individual risk estimation and such cases might represent a distinct, biologically determined disease subgroup.
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NOL3 antibody produced in rabbit, affinity isolated antibody