MilliporeSigma
  • Phase II study of cetuximab in combination with cisplatin and radiation in unresectable, locally advanced head and neck squamous cell carcinoma: Eastern cooperative oncology group trial E3303.
Phase II study of cetuximab in combination with cisplatin and radiation in unresectable, locally advanced head and neck squamous cell carcinoma: Eastern cooperative oncology group trial E3303.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-08-12)
Ann Marie Egloff, Ju-Whei Lee, Corey J Langer, Harry Quon, Alec Vaezi, Jennifer R Grandis, Raja R Seethala, Lin Wang, Dong M Shin, Athanassios Argiris, Donghua Yang, Ranee Mehra, John Andrew Ridge, Urjeet A Patel, Barbara A Burtness, Arlene A Forastiere
ABSTRACT
Treatment with cisplatin or cetuximab combined with radiotherapy each yield superior survival in locally advanced squamous cell head and neck cancer (LA-SCCHN) compared with radiotherapy alone. Eastern Cooperative Oncology Group Trial E3303 evaluated the triple combination. Patients with stage IV unresectable LA-SCCHN received a loading dose of cetuximab (400 mg/m(2)) followed by 250 mg/m(2)/week and cisplatin 75 mg/m(2) q 3 weeks ×3 cycles concurrent with standard fractionated radiotherapy. In the absence of disease progression or unacceptable toxicity, patients continued maintenance cetuximab for 6 to 12 months. Primary endpoint was 2-year progression-free survival (PFS). Patient tumor and blood correlates, including tumor human papillomavirus (HPV) status, were evaluated for association with survival. A total of 69 patients were enrolled; 60 proved eligible and received protocol treatment. Oropharyngeal primaries constituted the majority (66.7%), stage T4 48.3% and N2-3 91.7%. Median radiotherapy dose delivered was 70 Gy, 71.6% received all three cycles of cisplatin, and 74.6% received maintenance cetuximab. Median PFS was 19.4 months, 2-year PFS 47% [95% confidence interval (CI), 33%-61%]. Two-year overall survival (OS) was 66% (95% CI, 53%-77%); median OS was not reached. Response rate was 66.7%. Most common grade ≥3 toxicities included mucositis (55%), dysphagia (46%), and neutropenia (26%); one attributable grade 5 toxicity occurred. Only tumor HPV status was significantly associated with survival. HPV was evaluable in 29 tumors; 10 (all oropharyngeal) were HPV positive. HPV(+) patients had significantly longer OS and PFS (P = 0.004 and P = 0.036, respectively). Concurrent cetuximab, cisplatin, and radiotherapy were well tolerated and yielded promising 2-year PFS and OS in LA-SCCHN with improved survival for patients with HPV(+) tumors.
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin