MilliporeSigma
EphB6 promotes anoikis by modulating EphA2 signaling.
Cellular signalling (2014-09-23)
Mai Akada, Kohei Harada, Manabu Negishi, Hironori Katoh
ABSTRACT
Anoikis is a specific type of apoptosis induced by detachment of epithelial cells from extracellular matrix, and acquiring resistance to anoikis is an important step that enables cancer cells to metastasize. EphA2, which is overexpressed in a variety of human cancers, is phosphorylated by Akt on serine 897 and mediates ligand ephrin-independent promotion of anoikis resistance through the RhoG activator Ephexin4. EphB6 is frequently silenced in invasive and metastatic cancers; however, its role in cancer progression is poorly understood. Here we show that EphB6 interacts with EphA2 and suppresses EphA2-mediated promotion of anoikis resistance in MCF7 breast cancer cells. On the other hand, knockdown of EphB6 promotes anoikis resistance. We further show that expression of EphB6 decreases serine 897 phosphorylation of EphA2 and suppresses EphA2-Ephexin4 interaction and the RhoG activation. These findings implicate EphB6 as a negative regulator of EphA2 oncogenic signaling.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O