MilliporeSigma
  • Tirofiban induces VEGF production and stimulates migration and proliferation of endothelial cells.
Tirofiban induces VEGF production and stimulates migration and proliferation of endothelial cells.
Vascular pharmacology (2014-04-23)
Arturo Giordano, Anna D'Angelillo, Simona Romano, Paolo D'Arrigo, Nicola Corcione, Rita Bisogni, Stefano Messina, Michele Polimeno, Paolo Pepino, Paolo Ferraro, Maria Fiammetta Romano
ABSTRACT
Tirofiban is a fibrinogen receptor antagonist, generated using the tripeptide Arg-Gly-Asp (RGD) as a template. RGD activates integrin receptors and integrin-mediated signals are necessary for normal cells to promote survival and stimulate cell cycle progression. We investigated whether tirofiban activated growth-stimulatory signals in endothelium. For this study human umbilical vein endothelial cells (HUVEC) and human aortic endothelial cells (HAEC) were used. Analysis of cell proliferation, by cell counts, showed that the number of endothelial cells doubled after 72 h of culture in the absence of tirofiban, while they were tripled and even quadrupled, in the presence of increasing doses of the drug. Moreover, tirofiban-stimulated cells had a greater ability to migrate through the transwell filters of Boyden chamber, compared to unstimulated cells. The scratch assay, which mimics cell migration during wound healing, showed that tirofiban stimulated HUVECs to migrate into the leading hedge of the scratch. Western blot showed that tirofiban increased the expression levels of VEGF and the downstream effectors Erk and cyclin D. An inhibitor of VEGFR2 counteracted tirofiban-induced-proliferation, suggesting a role for VEGF in such effect. Our study shows that tirofiban stimulates the migration and proliferation of endothelial cells suggesting that it can promote endothelial repair. Ex vivo cultures of arterial rings confirmed the growth stimulatory effect of tirofiban on endothelium. Thus, the benefits of tirofiban in those with acute coronary syndromes undergoing PTCA may be due to rapid endothelization of damaged vessel, besides antiplatelet effects.
Sigma-Aldrich
Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Vascular Endothelial Growth Factor from mouse, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture