MilliporeSigma
  • Delayed noradrenergic activation in the dorsal hippocampus promotes the long-term persistence of extinguished fear.
Delayed noradrenergic activation in the dorsal hippocampus promotes the long-term persistence of extinguished fear.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2014-02-21)
Ning Chai, Jian-Feng Liu, Yan-Xue Xue, Chang Yang, Wei Yan, Hui-Min Wang, Yi-Xiao Luo, Hai-Shui Shi, Ji-Shi Wang, Yan-Ping Bao, Shi-Qiu Meng, Zeng-Bo Ding, Xue-Yi Wang, Lin Lu
ABSTRACT
Fear extinction has been extensively studied, but little is known about the molecular processes that underlie the persistence of extinction long-term memory (LTM). We found that microinfusion of norepinephrine (NE) into the CA1 area of the dorsal hippocampus during the early phase (0 h) after extinction enhanced extinction LTM at 2 and 14 days after extinction. Intra-CA1 infusion of NE during the late phase (12 h) after extinction selectively promoted extinction LTM at 14 days after extinction that was blocked by the β-receptor antagonist propranolol, protein kinase A (PKA) inhibitor Rp-cAMPS, and protein synthesis inhibitors anisomycin and emetine. The phosphorylation levels of PKA, cyclic adenosine monophosphate response element-binding protein (CREB), GluR1, and the membrane GluR1 level were increased by NE during the late phase after extinction that was also blocked by propranolol and Rp-cAMPS. These results suggest that the enhancement of extinction LTM persistence induced by NE requires the activation of the β-receptor/PKA/CREB signaling pathway and membrane GluR1 trafficking. Moreover, extinction increased the phosphorylation levels of Erk1/2, CREB, and GluR1, and the membrane GluR1 level during the late phase, and anisomycin/emetine alone disrupted the persistence of extinction LTM, indicating that the persistence of extinction LTM requires late-phase protein synthesis in the CA1. Propranolol and Rp-cAMPS did not completely disrupt the persistence of extinction LTM, suggesting that another β-receptor/PKA-independent mechanism underlies the persistence of extinction LTM. Altogether, our results showed that enhancing hippocampal noradrenergic activity during the late phase after extinction selectively promotes the persistence of extinction LTM.
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rp-Adenosine 3′,5′-cyclic monophosphorothioate triethylammonium salt, powder, ≥98% (HPLC)