MilliporeSigma
  • Effects of Na/K-ATPase and its ligands on bone marrow stromal cell differentiation.
Effects of Na/K-ATPase and its ligands on bone marrow stromal cell differentiation.
Stem cell research (2014-05-06)
Moustafa Sayed, Christopher A Drummond, Kaleigh L Evans, Steven T Haller, Jiang Liu, Zijian Xie, Jiang Tian
ABSTRACT
Endogenous ligands of Na/K-ATPase have been demonstrated to increase in kidney dysfunction and heart failure. It is also reported that Na/K-ATPase signaling function effects stem cell differentiation. This study evaluated whether Na/K-ATPase activation through its ligands and associated signaling functions affect bone marrow stromal cells (BMSCs, also known as bone marrow-derived mesenchymal stem cells) differentiation capacity. BMSCs were isolated from male Sprague-Dawley rats and cultured in minimal essential medium alpha (MEM-α) supplemented with 15% Fetal Bovine serum (FBS). The results showed that marinobufagenin (MBG), a specific Na/K-ATPase ligand, potentiated rosiglitazone-induced adipogenesis in these BMSCs. Meanwhile, it attenuated BMSC osteogenesis. Mechanistically, MBG increased CCAAT/enhancer binding protein alpha (C/EBPα) protein expression through activation of an extracellular regulated kinase (ERK) signaling pathway, which leads to enhanced rosiglitazone-induced adipogenesis. Inhibition of ERK activation by U0126 blocks the effect of MBG on C/EBPα expression and on rosiglitazone-induced adipogenesis. Reciprocally, MBG reduced runt-related transcription factor 2 (RunX2) expression, which resulted in the inhibition of osteogenesis induced by β-glycerophosphate/ascorbic acid. MBG also potentiated rosiglitazone-induced adipogenesis in 3T3-L1 cells and in mouse BMSCs. These results suggest that Na/K-ATPase and its signaling functions are involved in the regulation of BMSCs differentiation.