MilliporeSigma
  • Role of epicardial adipose tissue in coronary artery disease in non-obese patients.
Role of epicardial adipose tissue in coronary artery disease in non-obese patients.
Journal of cardiology (2013-11-16)
Tadateru Iwayama, Joji Nitobe, Tetsu Watanabe, Mitsunori Ishino, Harutoshi Tamura, Satoshi Nishiyama, Hiroki Takahashi, Takanori Arimoto, Tetsuro Shishido, Takehiko Miyashita, Takuya Miyamoto, Shuji Toyama, Mitsuaki Sadahiro, Isao Kubota
ABSTRACT
Epicardial adipose tissue (EAT) surrounding the heart may contribute to the development of coronary artery disease (CAD) through its local secretion of adipocytokines. Although the quantity of EAT is associated with obesity and metabolic syndrome, the role of EAT in the development of CAD in non-obese patients remains to be determined. This study included 41 patients with CAD who underwent coronary artery bypass graft surgery and 28 patients without CAD who underwent other cardiac surgery. EAT volume was measured by 64-slice multi-detector computed tomography before the surgery. We obtained pericardial fluid and epicardial and subcutaneous adipose tissue samples at the surgery. We investigated the relationship between EAT volume and adiponectin levels in pericardial fluid and incident CAD in patients with and without obesity (body mass index>25 kg/m(2)). There was no significant difference in EAT volume between obese patients with and without CAD (55.5 ± 40.2 mL vs. 40.1 ± 19.7 mL, p=0.323). However, EAT volume was significantly greater in non-obese patients with CAD compared to those without CAD (35.0 ± 18.8 mL vs. 15.7 ± 11.0 mL, p<0.001). Adiponectin concentrations in pericardial fluid were significantly lower in non-obese patients with CAD compared to those without CAD (2.7 ± 2.0 μg/mL vs. 4.3 ± 3.7 μg/mL, p=0.049), whereas the adiponectin levels were decreased in obese patients regardless of the presence of CAD. Non-obese patients with CAD had significantly larger size adipocytes in EAT but not subcutaneous adipose tissue compared to those without CAD. Multiple logistic regression analysis showed that increased EAT volume was independently associated with incident CAD in non-obese patients. Increased EAT may play a crucial role in development of CAD through impairment of adiponectin secretion in non-obese patients.
Product Description
Iopamidol, European Pharmacopoeia (EP) Reference Standard
USP
Iopamidol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet