MilliporeSigma
  • Up-regulation of HDAC4 is associated with Schwann cell proliferation after sciatic nerve crush.
Up-regulation of HDAC4 is associated with Schwann cell proliferation after sciatic nerve crush.
Neurochemical research (2014-08-12)
Yonghua Liu, Yang Liu, Xiaoke Nie, Jianhua Cao, Xiaojian Zhu, Weidong Zhang, Zhongbing Liu, Xingxing Mao, Shixian Yan, Yingjie Ni, Youhua Wang
ABSTRACT
Histone deacetylase 4 (HDAC4), a member of the class IIa HDACs subfamily, has emerged as a critical regulator of cell growth, differentiation, and migration in various cell types. It was reported that HDAC4 stimulated colon cell proliferation via repression of p21. Also, HDAC4 contributes to platelet-derived growth factor-BB-induced proliferation and migration of vascular smooth muscle cells. Furthermore, HDAC4 may play an important role in the regulation of neuronal differentiation and survival. However, the role of HDAC4 in the process of peripheral nervous system regeneration after injury remains virtually unknown. Herein, we investigated the spatiotemporal expression of HDAC4 in a rat sciatic nerve crush model. We found that sciatic nerve crush induced up-regulated expression of HDAC4 in Schwann cells. Moreover, the expression of the proliferation marker Ki-67 exhibited a similar tendency with that of HDAC4. In cell cultures, we observed increased expression of HDAC4 during the process of TNF-α-induced Schwann cell proliferation, whereas the protein level of p21 was down-regulated. Interference of HDAC4 led to enhanced expression of p21 and impaired proliferation of Schwan cells. Taken together, our findings implicated that HDAC4 was up-regulated in the sciatic nerve after crush, which was associated with proliferation of Schwann cells.
Product Description
USP
Cytarabine, United States Pharmacopeia (USP) Reference Standard
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC4
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution