- Association of OAZ1 gene polymorphisms with subclinical and clinical vascular events.
Association of OAZ1 gene polymorphisms with subclinical and clinical vascular events.
Proliferation and migration of vascular smooth muscle cells (VSMCs) are striking features shared by vascular ageing, atherosclerosis, and in-stent restenosis. VSMC biology depends in part on polyamines whose metabolism is closely regulated by ornithine decarboxylase antizyme 1 (OAZ1). Therefore, we sought for association between OAZ1 gene polymorphisms and various outcomes involving VSMC proliferation. Systematic screening of the OAZ1 gene enabled to detect 21 variants. The impact of 4 selected tag polymorphisms (+849C/T, +851G/T, +1804G/A, and +2222A/G) was evaluated in 3 independent association studies. In a sample of 205 patients, the +2222G allele was associated with an increased risk of 6-month coronary in-stent restenosis (OR [95%CI]=2.1 [1.2 to 3.6]; P=0.0071). In a sample of 1001 subjects participating to the EVA study, the +2222G allele was longitudinally associated with a 4-year increase in common carotid intima-media thickness (P=0.047). In a case-control study (466 cases versus 466 controls), the risk of coronary heart disease associated with the +2222G allele was 1.3 (95%CI=[1.1 to 1.6]; P=0.026). No other significant association was consistently detected. We identified the OAZ1+2222A/G polymorphism as a potential genetic marker of vascular events. Our findings strengthen the hypothesis that the polyamine metabolism plays a role in vascular diseases.