MilliporeSigma
  • Association of genetic variation of the RIL gene, encoding a PDZ-LIM domain protein and localized in 5q31.1, with low bone mineral density in adult Japanese women.
Association of genetic variation of the RIL gene, encoding a PDZ-LIM domain protein and localized in 5q31.1, with low bone mineral density in adult Japanese women.
Journal of human genetics (2003-08-09)
Fumihiro Omasu, Yoichi Ezura, Mitsuko Kajita, Ryota Ishida, Mina Kodaira, Hideo Yoshida, Takao Suzuki, Takayuki Hosoi, Satoshi Inoue, Masataka Shiraki, Hajime Orimo, Mitsuru Emi
ABSTRACT
Twin and family studies had shown that genetic factors are important determinants of bone mass. Multiple genes might be involved. One candidate gene, the reversion-induced LIM gene ( RIL), is a PDZ and LIM-domain-containing protein and has been localized within the cytokine cluster of chromosome 5 (5q31.1). In a genetic study of 370 adult Japanese women, we investigated the correlation between radial bone mineral density (BMD) and a genetic variation (-3333T-->C) of the 5'-flanking region of RIL gene. A significant association was identified between the RIL variation -3333T-->C and radial BMD ( r=0.15, P=0.003). The variation of the RIL locus may be an important determinant of osteoporosis.