MilliporeSigma
  • Mutations in the fatty acid transport protein 4 gene cause the ichthyosis prematurity syndrome.
Mutations in the fatty acid transport protein 4 gene cause the ichthyosis prematurity syndrome.
American journal of human genetics (2009-07-28)
Joakim Klar, Martina Schweiger, Robert Zimmerman, Rudolf Zechner, Hao Li, Hans Törmä, Anders Vahlquist, Bakar Bouadjar, Niklas Dahl, Judith Fischer
ABSTRACT
Ichthyosis prematurity syndrome (IPS) is an autosomal-recessive disorder characterized by premature birth and neonatal asphyxia, followed by a lifelong nonscaly ichthyosis with atopic manifestations. Here we show that the gene encoding the fatty acid transport protein 4 (FATP4) is mutated in individuals with IPS. Fibroblasts derived from a patient with IPS show reduced activity of very long-chain fatty acids (VLCFA)-CoA synthetase and a specific reduction in the incorporation of VLCFA into cellular lipids. The human phenotype is consistent with Fatp4 deficiency in mice that is characterized by a severe skin phenotype, a defective permeability barrier function, and perturbed VLCFA metabolism. Our results further emphasize the importance of fatty acid metabolism for normal epidermal barrier function illustrated by deficiency of a member in the FATP family of proteins.