MilliporeSigma
  • GPX2 overexpression is involved in cell proliferation and prognosis of castration-resistant prostate cancer.
GPX2 overexpression is involved in cell proliferation and prognosis of castration-resistant prostate cancer.
Carcinogenesis (2014-02-25)
Taku Naiki, Aya Naiki-Ito, Makoto Asamoto, Noriyasu Kawai, Keiichi Tozawa, Toshiki Etani, Shinya Sato, Shugo Suzuki, Tomoyuki Shirai, Kenjiro Kohri, Satoru Takahashi
ABSTRACT
There is a need for exploration of new therapeutic strategies that target distinct molecular mechanisms of castration-resistant prostate cancer (CRPC) because its emergence following androgen deprivation therapy is a major clinical problem. In this report, we investigated the role of glutathione peroxidase 2 (GPX2) in CRPC. GPX2 expression was analyzed in rat and human CRPC cells. Next, we determined the proliferation rate and level of reactive oxygen species (ROS) in GPX2-small interfering RNA (siRNA)-transfected CRPC cells. For in vivo analysis, siRNA-transfected cells were subcutaneously implanted into normal and castrated nude mice. Further, immunohistochemical and prognostic analyses of GPX2 were performed using human specimens. Silencing of GPX2 caused significant growth inhibition and increased intracellular ROS in both rat (PCai1) and human (PC3) CRPC cells. Flow cytometry and western blot analyses revealed that the decrease in proliferation rate of the GPX2-silenced cells was due to cyclin B1-dependent G2/M arrest. Furthermore, knockdown of Gpx2 inhibited tumor growth of PCai1 cells in castrated mice. Immunohistochemical analyses indicated that expression of GPX2 was significantly higher in residual cancer foci after neoadjuvant hormonal therapy than in hormone naive cancer foci. Moreover, patients with high GPX2 expression in biopsy specimen had significantly lower prostate-specific antigen recurrence-free survival and overall survival than those with no GPX2 expression. These findings suggest that GPX2 is a prognostic marker in CRPC and affects proliferation of prostate cancer under androgen depletion partially through protection against ROS signaling.
Product Description
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)