MilliporeSigma
EGFR signaling patterns are regulated by its different ligands.
Growth factors (Chur, Switzerland) (2014-09-27)
Stine Louise Jeppe Knudsen, Anni Sieu Wai Mac, Lasse Henriksen, Bo van Deurs, Lene Melsæther Grøvdal
ABSTRACT
EGF receptor (EGFR) and its signaling have been investigated for many years, but how its different ligands regulate signaling has not been thoroughly explored. When investigating EGFR activation and downstream signaling in HeLa cells using a panel of ligands, we found a ligand-dependent differential activation of EGFR and the signaling pathways Akt, PLCγ and STAT with HB-EGF and BTC being the most potent ligands. All the tested ligands induced full activation of Erk signaling at 1 nM, whereas only HB-EGF and partly BTC and EGF induced strong activation of Akt, STAT3 and PLCγ at this concentration. Interestingly, we also found that the high activation potencies of HB-EGF and BTC could only partially be explained by their binding affinities, and are therefore likely to be regulated by other mechanisms. We thus suggest that the signaling pathways initiated from the EGFR vary depending on the ligands bound in a cell specific manner.
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture