MilliporeSigma
  • Heparin/heparan sulfate controls fibrillin-1, -2 and -3 self-interactions in microfibril assembly.
Heparin/heparan sulfate controls fibrillin-1, -2 and -3 self-interactions in microfibril assembly.
FEBS letters (2014-07-19)
Laetitia Sabatier, Jelena Djokic, Dirk Hubmacher, Dzaner Dzafik, Valentin Nelea, Dieter P Reinhardt
ABSTRACT
Fibrillins form multifunctional microfibrils in most connective tissues. Deficiencies in fibrillin assembly can result in fibrillinopathies, such as Marfan syndrome. We demonstrate the presence of heparin/heparan sulfate binding sites in fibrillin-2 and -3. Multimerization of all three fibrillins drastically increased the apparent affinity of their interaction with heparin/heparan sulfate. Surprisingly, contrary to other reports heparin/heparan sulfate strongly inhibited homo- and heterotypic N-to-C-terminal fibrillin interactions. These data suggest that heparin/heparan sulfate controls the formation of microfibrils at the bead interaction stage.
Sigma-Aldrich
Monoclonal Anti-Fibronectin antibody produced in mouse, clone FN-15, ascites fluid
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg