MilliporeSigma
The "stop" and "go" of nicotine dependence: role of GABA and glutamate.
Cold Spring Harbor perspectives in medicine (2013-06-05)
Manoranjan S D'Souza, Athina Markou
ABSTRACT
Nicotine plays an important role in the initiation and maintenance of tobacco smoking. Importantly, chronic nicotine exposure alters the function of brain reward systems, resulting in the development of a nicotine-dependent state. This nicotine-dependent state is associated with aversive affective and somatic signs upon abstinence from smoking, often leading to relapse in abstinent smokers. This article reviews the role of the major excitatory and inhibitory neurotransmitters glutamate and γ-aminobutyric acid (GABA), respectively, in both the reinforcing effects of nicotine and development of nicotine dependence. Evidence suggests that blockade of glutamatergic neurotransmission attenuates both nicotine intake and nicotine seeking. In contrast, both nicotine intake and nicotine seeking are attenuated when GABA neurotransmission is facilitated. In conclusion, medications that either attenuate/negatively modulate glutamatergic neurotransmission or facilitate/positively modulate GABA neurotransmission may be useful for promoting smoking cessation in humans.
Product Description
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
S(−)-Nicotine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®