MilliporeSigma
  • Anaphylaxis to chlorhexidine-coated central venous catheters: a case series and review of the literature.
Anaphylaxis to chlorhexidine-coated central venous catheters: a case series and review of the literature.
Surgical infections (2012-05-10)
Achyut Guleri, Abhishek Kumar, Richard J M Morgan, Mike Hartley, David H Roberts
ABSTRACT
Anaphylactic reactions to chlorhexidine are rare but are being reported increasingly in association with a variety of products. We report three cases of anaphylaxis to chlorhexidine in patients presenting for cardiac surgery. In each case, anaphylaxis was precipitated by the insertion of a central venous catheter impregnated with chlorhexidine acetate. Subsequent investigations confirmed chlorhexidine as the causal agent. Extensive use of chlorhexidine to reduce hospital-acquired infections has the potential to sensitize a small proportion of patients, leading to life-threatening anaphylaxis on subsequent exposure.
Product Description
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorhexidine, ≥99.5%