MilliporeSigma
  • Determination of bromothalonil residues and degradation in apple and soil by QuEChERS and GC-MS/MS.
Determination of bromothalonil residues and degradation in apple and soil by QuEChERS and GC-MS/MS.
Bulletin of environmental contamination and toxicology (2014-02-20)
Huijun Liu, Baoyuan Guo, Huili Wang, Jianzhong Li, Lin Zheng
ABSTRACT
The dissipation and residues of bromothalonil in apple and soil under field condition were analyzed by QuEChERS (quick, easy, cheap, effective, rugged and safe) combined with gas chromatography-mass spectrometer method. The recoveries were ranged from 80.8 % to 106 % with coefficient variation for repeatability ranged from 3.08 % to 7.09 % at fortification levels of 0.02, 0.04 and 0.2 mg/kg in apple and soil. The limit of quantification of the method was 0.31 μg/kg. The dissipation rates of bromothalonil followed the first-order kinetics and the half-lives were from 3.61 to 3.98 days in apple and from 4.65 to 9.29 days in soil. In apple, the terminal residues of bromothalonil were below the China maximum residue limit (0.2 mg/kg) after 7 days of application. This work contributed to provide the basic information for a safe usage of bromothalonil in apple orchard and preventing health problem from consumers in China.
Product Description
Supelco
Methyldibromoglutaronitrile, analytical standard