MilliporeSigma
  • Validation of suitable reference genes for expression studies in different pilocarpine-induced models of mesial temporal lobe epilepsy.
Validation of suitable reference genes for expression studies in different pilocarpine-induced models of mesial temporal lobe epilepsy.
PloS one (2013-09-07)
Thalita Ewellyn Batista Sales Marques, Leila Rodrigues de Mendonça, Marília Gabriela Pereira, Tiago Gomes de Andrade, Norberto Garcia-Cairasco, Maria Luisa Paçó-Larson, Daniel Leite Góes Gitaí
ABSTRACT
It is well recognized that the reference gene in a RT-qPCR should be properly validated to ensure that gene expression is unaffected by the experimental condition. We investigated eight potential reference genes in two different pilocarpine PILO-models of mesial temporal lobe epilepsy (MTLE) performing a stability expression analysis using geNorm, NormFinder and BestKepeer softwares. Then, as a validation strategy, we conducted a relative expression analysis of the Gfap gene. Our results indicate that in the systemic PILO-model Actb, Gapdh, Rplp1, Tubb2a and Polr1a mRNAs were highly stable in hippocampus of rats from all experimental and control groups, whereas Gusb revealed to be the most variable one. In fact, we observed that using Gusb for normalization, the relative mRNA levels of the Gfap gene differed from those obtained with stable genes. On the contrary, in the intrahippocampal PILO-model, all softwares included Gusb as a stable gene, whereas B2m was indicated as the worst candidate gene. The results obtained for the other reference genes were comparable to those observed for the systemic Pilo-model. The validation of these data by the analysis of the relative expression of Gfap showed that the upregulation of the Gfap gene in the hippocampus of rats sacrificed 24 hours after status epilepticus (SE) was undetected only when B2m was used as the normalizer. These findings emphasize that a gene that is stable in one pathology model may not be stable in a different experimental condition related to the same pathology and therefore, the choice of reference genes depends on study design.