MilliporeSigma
  • Down-regulation of TSG101 by small interfering RNA inhibits the proliferation of breast cancer cells through the MAPK/ERK signal pathway.
Down-regulation of TSG101 by small interfering RNA inhibits the proliferation of breast cancer cells through the MAPK/ERK signal pathway.
Histology and histopathology (2010-12-01)
Y Zhang, M Song, Z S Cui, C Y Li, X X Xue, M Yu, Y Lu, S Y Zhang, E H Wang, Y Y Wen
ABSTRACT
We designed to investigate the effects of down-regulating the tumor susceptibility gene 101 (TSG101) on the proliferation and apoptosis of the human breast cancer MCF-7 cell line, and the role of the MAPK/ERK signal pathway in this process. The siRNA against TSG101 was transfected into the breast cancer MCF-7 cell line using Lipofectamine 2000. After TSG101 knockdown, the proliferation of MCF-7 cells was measured by the MTT assay. The cell cycle distribution and apoptosis were examined by using flow cytometry while cell migration was measured using a transwell assay. The protein level of p-ERK was further assessed by immunofluorescence and western blotting. Our results are as following, the MCF-7 cells transfected with TSG101 siRNA proliferated significantly slower and exhibited significantly increased rates of apoptosis compared to the control cells. In the TSG101 siRNA transfected cells, the percentage of cells in the G₀/G₁ and S phase of the cell cycle was significantly higher and lower, respectively, compared to the control cells. Moreover, the migration ability of TSG101 siRNA transfected cells was lower than the control groups. Lastly, the level of p-ERK protein in TSG101 siRNA transfected cells was significantly decreased compared with the control cells. In conclusion, TSG101 knockdown in breast cancer cells induces apoptosis and inhibits proliferation. The TSG101 depleted cells are arrested at the G₁/S transition of the cell cycle. The migration of breast cancer cells is also impaired by TSG101 siRNA. TSG101 may play a biological role through modulation of the MAPK/ERK signaling pathway in breast cancer.