MilliporeSigma
  • A compact hybrid-multiplexed potentiostat for real-time electrochemical biosensing applications.
A compact hybrid-multiplexed potentiostat for real-time electrochemical biosensing applications.
Biosensors & bioelectronics (2013-04-30)
Ioannis Ramfos, Nikolaos Vassiliadis, Spyridon Blionas, Konstantinos Efstathiou, Alex Fragoso, Ciara K O'Sullivan, Alexios Birbas
ABSTRACT
The architecture and design of a compact, multichannel, hybrid-multiplexed potentiostat for performing electrochemical measurements on continuously-biased electrode arrays is presented. The proposed architecture utilises a combination of sequential and parallel measurements, to enable high performance whilst keeping the system low-cost and compact. The accuracy of the signal readout is maintained by following a special multiplexing approach, which ensures the continuous biasing of all the working electrodes of an array. After sampling the results, a digital calibration technique factors out errors from component inaccuracies. A prototype printed circuit board (PCB) was designed and built using off-the-shelf components for the real-time measurement of the amperometric signal of 48 electrodes. The operation and performance of the PCB was evaluated and characterised through a wide range of testing conditions, where it exhibited high linearity (R(2)>0.999) and a resolution of 400pA. The effectiveness of the proposed multiplexing scheme is demonstrated through electrochemical tests using KCl and [Fe(CN)6](3-) in KCl solutions. The applicability of the prototype multichannel potentiostat is also demonstrated using real biosensors, which were applied to the detection of IgA antibodies.
Sigma-Aldrich
Potassium chloride, ≥99.0%
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride, BioUltra, ≥99.5% (AT)
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
Potassium standard solution, suitable for atomic absorption spectrometry, 1000 ppm K+, 1 mg/mL K+
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, ~1 M in H2O
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)