MilliporeSigma
  • Assessment of SOX11 expression in routine lymphoma tissue sections: characterization of new monoclonal antibodies for diagnosis of mantle cell lymphoma.
Assessment of SOX11 expression in routine lymphoma tissue sections: characterization of new monoclonal antibodies for diagnosis of mantle cell lymphoma.
The American journal of surgical pathology (2013-10-23)
Davide Soldini, Alexandra Valera, Carla Solé, Jara Palomero, Virginia Amador, José Ignacio Martin-Subero, Inmaculada Ribera-Cortada, Cristina Royo, Itziar Salaverria, Sílvia Beà, Elena Gonzalvo, Henrik Johannesson, Mileyka Herrera, Lluís Colomo, Antonio Martinez, Elias Campo
ABSTRACT
The diagnosis of mantle cell lymphoma (MCL) can be difficult, especially when no t(11;14) translocation and cyclin D1 overexpression can be detected. In such cases, the transcription factor SOX11 represents an important diagnostic marker, as it is expressed in most MCLs and, in particular, in all cyclin D1-negative MCLs reported so far. A reliable anti-SOX11 antibody is therefore a very useful tool for routine diagnosis. Here, we characterize the new monoclonal anti-SOX11 antibodies, suitable for Western blot assay and immunohistochemistry (IHC) on formalin-fixed paraffin-embedded tissue; we tested them on a large series of primary lymphoid tumors and compared these results with those of other routinely used antibodies. Moreover, we show that IHC results depend on transcription levels of SOX11, which suggests that posttranscriptional and posttranslational modifications do not significantly affect cutoff levels for IHC detection of SOX11.
Sigma-Aldrich
Anti-SOX11 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution