- Triptolide directly inhibits dCTP pyrophosphatase.
Triptolide directly inhibits dCTP pyrophosphatase.
Chembiochem : a European journal of chemical biology (2011-06-15)
Timothy W Corson, Hüseyin Cavga, Nicholas Aberle, Craig M Crews
PMID21671327
ABSTRACT
Triptolide is a potent natural product, with documented antiproliferative, immunosuppressive, anti-inflammatory, antifertility, and antipolycystic kidney disease effects. Despite a wealth of knowledge about the biology of this compound, direct intracellular target proteins have remained elusive. We synthesized a biotinylated photoaffinity derivative of triptolide, and used it to identify dCTP pyrophosphatase 1 (DCTPP1) as a triptolide-interacting protein. Free triptolide interacts directly with recombinant DCTPP1, and inhibits the enzymatic activity of this protein. Triptolide is thus the first dCTP pyrophosphatase inhibitor identified, and DCTPP1 is a biophysically validated target of triptolide.