MilliporeSigma
  • MK2 targets AU-rich elements and regulates biosynthesis of tumor necrosis factor and interleukin-6 independently at different post-transcriptional levels.
MK2 targets AU-rich elements and regulates biosynthesis of tumor necrosis factor and interleukin-6 independently at different post-transcriptional levels.
The Journal of biological chemistry (2001-12-14)
Armin Neininger, Dimitris Kontoyiannis, Alexey Kotlyarov, Reinhard Winzen, Rolf Eckert, Hans-Dieter Volk, Helmut Holtmann, George Kollias, Matthias Gaestel
ABSTRACT
We demonstrate that lipopolysaccharide-induced tumor necrosis factor (TNF) biosynthesis becomes independent of MAPKAP kinase 2 (MK2) when the AU-rich element (ARE) of the TNF gene is deleted. In spleen cells and macrophages where TNF biosynthesis is restored as a result of this deletion, interleukin (IL)-6 biosynthesis is still dependent on MK2. In MK2-deficient macrophages the half-life of IL-6 mRNA is reduced more than 10-fold, whereas the half-life of TNF mRNA is only weakly decreased. It is shown that the stability of a reporter mRNA carrying the AU-rich 3'-untranslated region (3'-UTR) of IL-6 is increased by MK2. The data provide in vivo evidence that the AU-rich 3'-UTRs of TNF and IL-6 are downstream to MK2 signaling and make MK2 an essential component of mechanisms that regulate biosynthesis of IL-6 at the levels of mRNA stability, and of TNF mainly through TNF-ARE-dependent translational control.
Sigma-Aldrich
Lipopolysaccharides from Salmonella typhosa, γ-irradiated, BioXtra, suitable for cell culture