MilliporeSigma
  • Interactions between modulators of the GABA(A) receptor: Stiripentol and benzodiazepines.
Interactions between modulators of the GABA(A) receptor: Stiripentol and benzodiazepines.
European journal of pharmacology (2011-01-18)
Janet L Fisher
ABSTRACT
Many patients with refractory epilepsy are treated with polytherapy, and nearly 15% of epilepsy patients receive two or more anti-convulsant agents. The anti-convulsant stiripentol is used as an add-on treatment for the childhood epilepsy syndrome known as severe myoclonic epilepsy in infancy (Dravet syndrome). Stiripentol has multiple mechanisms of action, both enhancing GABA(A) receptors and reducing activity of metabolic enzymes that break down other drugs. Stiripentol is typically co-administered with other anti-convulsants such as benzodiazepines which also act through GABA(A) receptor modulation. Stiripentol slows the metabolism of some of these drugs through inhibition of a variety of cytochrome P450 enzymes, but could also influence their effects on GABAergic neurotransmission. Is it rational to co-administer drugs which can act through the same target? To examine the potential interaction between these modulators, we transiently transfected HEK-293T cells to produce α3β3γ2L or α3β3δ recombinant GABA(A) receptors. Using whole-cell patch clamp recordings, we measured the response to each benzodiazepine alone and in combination with a maximally effective concentration of stiripentol. We compared the responses to four different benzodiazepines: diazepam, clonazepam, clobazam and norclobazam. In all cases we found that these modulators were equally effective in the presence and absence of stiripentol. The δ-containing receptors were insensitive to modulation by the benzodiazepines, which did not affect potentiation by stiripentol. These data suggest that stiripentol and the benzodiazepines act independently at GABA(A) receptors and that polytherapy could be expected to increase the maximum effect beyond either drug alone, even without consideration of changes in metabolism.
Sigma-Aldrich
Stiripentol, ≥98% (HPLC)