MilliporeSigma
Child health update: estrogen cream for labial adhesion in girls.
Canadian family physician Medecin de famille canadien (2013-01-24)
Ran D Goldman
ABSTRACT
Should I prescribe estrogen cream to treat labial adhesion in my prepubertal female patients? Many physicians offer young girls estrogen cream to be applied on the labia for several weeks to treat labial adhesion. While no randomized controlled trial data are available, the success rate of such creams is reported in the literature to be close to 90%. Side effects of estrogen cream are mild and transient. Although the ideal frequency and length of treatment with estrogen cream has yet to be determined, most treatment recommendations suggest application of the cream 1 to 2 times a day; treatment should be applied for a few weeks before considering surgical alternatives.
Product Description
Supelco
Betamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Betamethasone, ≥98%
Betamethasone, European Pharmacopoeia (EP) Reference Standard