MilliporeSigma
  • High serum levels of free cortisol indicate severity of cirrhosis in hemodynamically stable patients.
High serum levels of free cortisol indicate severity of cirrhosis in hemodynamically stable patients.
Journal of gastroenterology and hepatology (2012-06-01)
Thierry Thevenot, Richard Dorin, Elisabeth Monnet, Clifford R Qualls, Remy Sapin, Emilie Grandclement, Sophie Borot, Frances Sheppard, Delphine Weil, Thibault Degand, Vincent Di Martino, Rasa Kazlauskaite
ABSTRACT
We investigated: (i) the association between severity of cirrhosis and serum levels of free cortisol (SFC) and total cortisol (STC), measured before and 30 min after (T(30)) the low-dose 1-µg short synacthen test (LD-SST); and (ii) the prognostic value of SFC and STC. Consecutive, hemodynamically stable, cirrhotic patients (34 Child-Pugh class A, 29B, and 32C) underwent the LD-SST. Patients were followed for at least 12 months to assess non-transplant-related mortality. Child-Pugh class C patients had significantly higher basal levels of SFC than Child-Pugh class A or B patients. Prevalence of suspected adrenal dysfunction ranged between 7.4% (T(0) STC < 138 nmol/L) and 49.4% (change in STC < 250 nmol/L) according to the threshold used. In receiver-operator curve analysis, the area-under-the-curve values were 0.67 for T(30) SFC (0.51-0.79), 0.81 for Child-Pugh score (0.70-0.88), and 0.79 for albumin level (0.63-0.88). During the follow-up period, 16 patients with high T(30) SFC (≥ 78.9 nmol/L) (26.2%) and one patient with low T(30) SFC (< 78.9 nmol/L) (3.4%) died (P = 0.027 for high vs low T(30) SFC, log-rank test). Albeit not statistically significant, the risk of death for patients with T(30) SFC ≥ 78.9 nmol/L was fivefold higher than for patients with lower levels after adjusting for cirrhosis severity and level of albumin. One-year, non-transplant-related mortality is high among patients with T(30) levels of SFC ≥ 78.9 nmol/L (26.2%). These findings might result from latent inflammatory stress in hemodynamically stable cirrhotic patients, detected by adrenal testing.
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 1-24 human, rat, ≥97% (HPLC)