MilliporeSigma
  • Meldrum's acids and 5-alkylidene Meldrum's acids in catalytic carbon-carbon bond-forming processes.
Meldrum's acids and 5-alkylidene Meldrum's acids in catalytic carbon-carbon bond-forming processes.
Accounts of chemical research (2009-12-17)
Aaron M Dumas, Eric Fillion
ABSTRACT
Meldrum's acid (2,2-dimethyl-1,3-dioxane-4,6-dione) is a molecule with a unique history, owing to its originally misassigned structure, as well as a unique place among acylating agents, owing to its high acidity and remarkable electrophilicity. In this Account, we outline the work of our group and others toward harnessing the reactivity of Meldrum's acid derivatives in catalytic C-C bond-forming reactions. Taking advantage of the ability of Meldrum's acid to decompose to CO(2) and acetone following acyl substitution, we have shown that intramolecular Friedel-Crafts acylations can be performed under mild Lewis acidic conditions to yield a variety of benzocyclic ketones. In a further expansion of this method, a domino Friedel-Crafts acylation/alpha-tert alkylation reaction was used to complete the first total synthesis of (+/-)-taiwaniaquinol B. The unique characteristics of Meldrum's acid extend to its alkylidene derivatives, which have also proven exceptionally useful for the development of new reactions not readily accessible from other unsaturated carbonyl electrophiles. By combining the electrophilicity and dienophilicity of alkylidene Meldrum's acid with our Friedel-Crafts chemistry, we have demonstrated new domino syntheses of coumarin derivatives and tetrahydrofluorenones by conjugate additions, Diels-Alder cycloadditions, and C-H functionalizations. Additionally, we have used these powerful acceptors to allow conjugate alkenylation with functionalized organostannanes, and conjugate allylation under very mild conditions. We have also shown that these molecules permit the asymmetric formation of all-carbon quaternary stereocenters via enantioselective conjugate additions. These reactions employ dialkylzinc nucleophiles, maximizing functional group compatibility, while the presence of a Meldrum's acid moiety in the product allows a variety of postaddition modifications. A full investigation of this reaction has determined the structural factors of the alkylidene that contribute to optimal enantioselectivity. We have also used these acceptors to form tertiary propargylic stereocenters in very high enantiomeric excess by an extremely mild, Rh(I)-catalyzed addition of TMS-acetylene. Overall, we demonstrate that Meldrum's acid and its derivatives provide access to a broad range of reactivities that, combined with their ease of handling and preparation, make them ideal electrophiles.
Sigma-Aldrich
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%